Learn More
VU 0357017 hydrochloride, Tocris Bioscience™
Positive allosteric modulator of M1 receptors
Marke:  RD Systems (Bio-Techne) 4295/50
Weitere Details : Netto-Gewicht : 0.00050kg
Menge. 50mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13414514
  • 721.88 € / 50 Milligramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Positive allosteric modulator of muscarinic M 1 receptors (EC 50 = 198 nM). Displays no activity at M 2 -M 5 at concentrations up to 30 μM. Potentiates NMDA receptor currents in hippocampal neurons; activity reverses cognitive decifits in a rodent model of hippocampal-dependent memory. CNS penetrant.
Spezifikation
Chemische Identifikatoren
1135242-13-5
369.89
vu 0357017 hydrochloride, vu0357017 hydrochloride, vu0357017 hydrochloride, ethyl 4-2-2-methylbenzamido ethylamino piperidine-1-carboxylate hydrochloride, 4-2-2-methylbenzoyl amino ethyl amino-1-piperidinecarboxylic acid ethyl ester hydrochloride, ml071 hydrochloride, vu0357017 monohydrochloride, cid, hcl, vu0357017 monohydrochloride hplc
ethyl 4-[2-[(2-methylbenzoyl)amino]ethylamino]piperidine-1-carboxylate;hydrochloride
C18H28ClN3O3
XKJQVUIXSBOCPP-UHFFFAOYSA-N
25010775
CCOC(=O)N1CCC(CC1)NCCNC(=O)C2=CC=CC=C2C.Cl
Spezifikation
VU 0357017 hydrochloride
1135242-13-5
vu 0357017 hydrochloride, vu0357017 hydrochloride, vu0357017 hydrochloride, ethyl 4-2-2-methylbenzamido ethylamino piperidine-1-carboxylate hydrochloride, 4-2-2-methylbenzoyl amino ethyl amino-1-piperidinecarboxylic acid ethyl ester hydrochloride, ml071 hydrochloride, vu0357017 monohydrochloride, cid, hcl, vu0357017 monohydrochloride hplc
CCOC(=O)N1CCC(CC1)NCCNC(=O)C2=CC=CC=C2C.Cl
369.89
369.89
50mg
C18H28ClN3O3
XKJQVUIXSBOCPP-UHFFFAOYSA-N
ethyl 4-[2-[(2-methylbenzoyl)amino]ethylamino]piperidine-1-carboxylate;hydrochloride
25010775
>98%
Videos