Learn More
Fisherbrand™ Weiße Verschlusskappe aus Polypropylen mit Dichtungseinlage aus PE-Schaum
Catalog Number 11528202
Click to view available options
28mm-400 screw cap size; 4300/Cs.
Offer good chemical resistance. Fisherbrand™ White Polypropylene PE Foam Caps are known for good impact strength and pliability.
Resistant against acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Verschlussauskleidung Polyethylene Foam
Verschlussmaterial Plastic
Verschlussgröße 28-400 mm
Verschlusstyp Screw Cap
Produkttyp PE Foam Lined Cap
Farbe White
Beschreibung Screw cap
Material Polypropylene
Menge 4300/Cs.
Halsausführung 28-400
Form Round