missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRAP53 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14761-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.Spezifikation
WRAP53 | |
Polyclonal | |
Unconjugated | |
WRAP53 | |
TCAB1, WDR79, WRAP53 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216853, 287432, 55135 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XII5, Q8VC51, Q9BUR4 | |
Wrap53 | |
WRAP53 Fusion Protein Ag6522 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |