Learn More
Tocris Bioscience™ WWL 113
Ces3 and Ces1f inhibitor
N.V.
Spezifikation
Produkttyp | WWL 113 |
---|---|
Beschreibung | WWL 113 |
Molekulargewicht | 466.53 |
Beschreibung
Carboxylesterase Ces3 and Ces1f inhibitor (IC50 values are ∽100 nM). Also inhibits ABHD6. Exhibits minimal activity against a panel of other serine hydrolases. Promotes lipid storage in adipocytes. Reduces weight gain in diet-induced obese mice and enhances glucose tolerance in diabetic mice. Orally active.Spezifikation
WWL 113 | |
WWL 113 | |
466.53 |
For Research Use Only