missing translation for 'onlineSavingsMsg'
Learn More

YBEY Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA553745

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15741681

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: MSLVIRNLQR VIPIRRAPLR SKIEIVRRIL GVQKFDLGII CVDNKNIQHI NRIYRDRNVP TDVLSFPFHE HLKAGEFP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.

YBEY Polyclonal antibody specifically detects YBEY in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

YBEY
Polyclonal
Unconjugated
YBEY
1110068N23Rik; A130042E20Rik; C21orf57; Endoribonuclease YbeY; ORF57; ORF66; putative metalloprotease C21orf57; putative metalloprotease C21orf57 homolog; putative ribonuclease; RGD1308722; rRNA maturation factor homolog; YBEY; ybeY metallopeptidase; ybeY metallopeptidase (putative)
Rabbit
Antigen affinity chromatography
RUO
216119, 361822, 54059
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P58557, Q8CAV0
YBEY
Recombinant protein corresponding to Human YBEY
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only