Learn More
YIF1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563100
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HQLFDDTSSA QSRGYGAQRA PGGLSYPAAS PTPHAAFLAD PVSNMAMAYG SSLAAQGKEL VDKNIDRFIP ITK Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
YIF1B Polyclonal antibody specifically detects YIF1B in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
YIF1B | |
Polyclonal | |
Unconjugated | |
YIF1B | |
9430029K10Rik; FinGER8; PP4519; Protein YIF1B; UNQ3073/PRO9905; YIF1B; Yip1 interacting factor homolog B; Yip1 interacting factor homolog B (S. cerevisiae); Yip1 interacting factor homolog B, membrane trafficking protein; Yip1b; YIP1-interacting factor homolog B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90522 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5BJH7 | |
YIF1B | |
Recombinant protein corresponding to Human YIF1B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only