Learn More
YIPF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553656
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PTNGDFTFVS SADAEDLSGS IASPDVKLNL GGDFIKESTA TTFLRQRGYG WLLEVEDDDP EDNKPLLEEL DIDLKDIYYK IRCVLMPMPS LGFN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
YIPF4 Polyclonal antibody specifically detects YIPF4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
YIPF4 | |
Polyclonal | |
Unconjugated | |
YIPF4 | |
2310034L04Rik; FinGER4; Nbla11189; Protein YIPF4; putative protein product of Nbla11189; RGD1310157; Yip1 domain family member 4; Yip1 domain family, member 4; YIP1 family member 4; Yipf4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BSR8 | |
YIPF4 | |
Recombinant protein corresponding to Human YIPF4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only