Learn More
YIPF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551990
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: AGLSDISISQ DIPVEGEITI PMRSRIREFD SSTLNESVRN TIMRDL Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.
YIPF6 Polyclonal antibody specifically detects YIPF6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
YIPF6 | |
Polyclonal | |
Unconjugated | |
YIPF6 | |
A430107J06Rik; FinGER6; Protein YIPF6; Yip1 domain family member 6; Yip1 domain family, member 6; YIP1 family member 6; yipf6; zgc:86904 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EC8 | |
YIPF6 | |
Recombinant protein corresponding to Human YIPF6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only