Learn More
YJEFN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563901
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SGWDAETGSD SEDGLRPDVL VSLAAPKRCA GRFSGRHHFV AGRFVPDDVR RKFALRLPGY TGTD Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
YJEFN3 Polyclonal antibody specifically detects YJEFN3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
YJEFN3 | |
Polyclonal | |
Unconjugated | |
YJEFN3 | |
apolipoprotein A1 binding protein; hYjeF_N3; hYjeF_N3-19p13.11; YjeF N-terminal domain containing 3; yjeF N-terminal domain-containing protein 3; YjeF_N3; YJEFN3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374887 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6XGL0 | |
YJEFN3 | |
Recombinant protein corresponding to Human YJEFN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |