missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YOD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550157
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total YOD1 protein.
YOD1 Polyclonal antibody specifically detects YOD1 in Human, Rat samples. It is validated for Western Blot.Spezifikation
YOD1 | |
Polyclonal | |
Unconjugated | |
YOD1 | |
9930028C20Rik; DUBA8; DUBA-8; fe01a10; HIN7; HIN-7; HIV-1-induced protease 7; HIV-induced protein-7-like protease; hsHIN7; hypothetical protein LOC550411; OTU domain containing 2; OTU domain-containing protein 2; OTUD2; PRO0907; Ubiquitin thioesterase OTU1; wu:fe01a10; Yod1; YOD1 deubiquitinase; YOD1 OTU deubiquinating enzyme 1 homolog; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast); YOD1 OTU deubiquitinating enzyme 1 homologue; YOD1 OTU deubiquitinating enzyme 1 homologue (S. cerevisiae); zgc:112182 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
363982, 55432 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q32Q05, Q5VVQ6 | |
YOD1 | |
Synthesized peptide derived from internal of human YOD1. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |