missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YPEL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567583
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with Mouse and Rat samples.
YPEL4 Polyclonal antibody specifically detects YPEL4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
YPEL4 | |
Polyclonal | |
Unconjugated | |
YPEL4 | |
Protein yippee-like 4; yippee like 4; yippee-like 4; yippee-like 4 (Drosophila); YPEL4 | |
Rabbit | |
affinity chromatography | |
RUO | |
219539, 241525, 502643 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q5XID5, Q65Z93, Q96NS1 | |
YPEL4 | |
Synthesized peptide derived from human YPEL4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |