missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHDF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA549875
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total YTHDF1 protein.
YTHDF1 Polyclonal antibody specifically detects YTHDF1 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
YTHDF1 | |
Polyclonal | |
Unconjugated | |
YTHDF1 | |
2210410K23Rik; 8030473O16; C20orf21; DACA-1; DACA-1 homolog; Dermatomyositis associated with cancer putative autoantigen 1; dermatomyositis associated with cancer putative autoantigen 1 homolog; fb49c02; wu:fb49c02; wu:fi19h06; wu:fj87f09; wu:fp06c11; YTH domain family 1; YTH domain family protein 1; YTH domain family, member 1; YTH domain-containing family protein 1; YTH N(6)-methyladenosine RNA binding protein 1; YTH N6-methyladenosine RNA binding protein 1; Ythdf1; zgc:55657 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228994, 296467, 54915 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P59326, Q9BYJ9 | |
YTHDF1 | |
Synthesized peptide derived from N-terminal of human YTHDF1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |