missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YY1AP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10425-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The encoded gene product presumably interacts with YY1 protein; however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
YY1AP1 | |
Polyclonal | |
Unconjugated | |
YY1AP1 | |
HCCA1, HCCA2, YAP, YY1 associated protein 1, YY1AP, YY1AP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55249 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H869 | |
YY1AP1 | |
YY1AP1 Fusion Protein Ag0692 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |