missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB48 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24665-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ZBTB48 contains 1 BTB (POZ) domain and 11 C2H2-type zinc fingers. It belongs to the krueppel C2H2-type zinc-finger protein family and binds to and regulates the J and/or S elements in MHC II promoter.Spezifikation
ZBTB48 | |
Polyclonal | |
Unconjugated | |
ZBTB48 | |
HKR3, pp9964, Protein HKR3, TZAP, ZBTB48, ZNF855 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3104 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10074 | |
ZBTB48 | |
ZBTB48 Fusion Protein Ag20301 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |