missing translation for 'onlineSavingsMsg'
Learn More

Zinkchlorid, 1 M in Diethylether, Thermo Scientific Chemicals

Catalog Number p-7049728
Click to view available options
Menge:
100 mL
500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
Dichlorozink
Cl[Zn]Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
Dichlorozink
Cl[Zn]Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Zinc chloride
Farbe Farblos
Physikalische Form Flüssig
Menge 100 mL
Strukturformel ZnCl2
UN-Nummer UN2924
Flammpunkt -40°C (-40°F)
Merck Index 14,10132
Formelmasse 136.29
Concentration or Composition (by Analyte or Components) 1M in diethyl ether
Empfindlichkeit Hygroscopic
Dichte 0.835 g/mL
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'