Learn More
Zinc-Copper Couple, TRC
Product Code. 30450587
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450587 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30450587 Supplier Toronto Research Chemicals Supplier No. TRCZ4390001G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Zinc-Copper Couple
CAS 53801-63-1
Molecular Formula CuZn
Quantity 1 g
Synonym Copper','compd. with zinc
InChI Formula InChI=1S/Cu.Zn
SMILES [Cu].[Zn]
IUPAC Name copper;zinc
Molecular Weight (g/mol) 128.93
Formula Weight 126.8587