missing translation for 'onlineSavingsMsg'
Learn More

Zinkfolie, 0.62 mm (0.024 Zoll) Dicke, 30 cm (12 Zoll) Breite, 99.9 % (Metallbasis), Thermo Scientific Chemicals

Catalog Number p-4825004
Click to view available options
Menge:
50 cm
250 cm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
Zink
[Zn]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
Zink
[Zn]

Form: Ca. .665 g/30 x 50 cm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Zinc
Farbe Grau
Aussehen Gray foil
Prozentgehaltsbereich 99.9% (Metals basis)
Menge 50 cm
Geruch Odorless
Form Folie, 0.62 mm (0.024 Zoll) dick, 30 cm (12 Zoll) breit

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'