missing translation for 'onlineSavingsMsg'
Learn More

Zink-Ingot, 345 mm (13.6 Zoll) Länge, 99.99 % (Metallbasis), Thermo Scientific Chemicals

Catalog Number p-7048242
Click to view available options
Menge:
1 Pc.
5 Pc.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
Zink
[Zn]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
Zink
[Zn]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Zinc ingot
Namenshinweis 345mm (13.6 in.) long
Hinweise zum Prozentgehalt (Metallbasis)
Menge 1 Pc.
Reinheit (%) 99.99%
Geruch Odorless
Gewicht ≈175g/ingot
Form Ignot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'