missing translation for 'onlineSavingsMsg'
Learn More

Zinterol hydrochloride, Tocris Bioscience™

Selective β2 agonist

Marke:  Tocris (Bio-Techne) 105150

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418393

  • 916.43 € / 50 Milligramm


Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Potent and selective β2-adrenoceptor agonist (pKB values are 8.3 and < 5.7 for β 2 and β1 receptors respectively, as measured in human artery).
Spezifikation

Chemische Identifikatoren

37000-20-7
378.487
zinterol, zinterol inn, zinterolum inn-latin, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-ylamino ethyl phenyl methanesulfonamide, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-yl amino ethyl phenyl methanesulfonamide, zinterolum, 2'-hydroxy-5'-l-hydroxy-2-2-methyl-1-phenyl-2-propylamino ethyl methanesulfonanilide, methanesulfonamide, n-5-2-1,1-dimethyl-2-phenylethyl amino-1-hydroxyethyl-2-hydroxyphenyl, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-yl amino ethyl phenyl methanesulfonamide, d0a0kp
N-[2-hydroxy-5-[1-hydroxy-2-[(2-methyl-1-phenylpropan-2-yl)amino]ethyl]phenyl]methanesulfonamide
C19H26N2O4S
XJBCFFLVLOPYBV-UHFFFAOYSA-N
37990
CC(C)(CC1=CC=CC=C1)NCC(C2=CC(=C(C=C2)O)NS(=O)(=O)C)O

Spezifikation

Zinterol hydrochloride
37000-20-7
zinterol, zinterol inn, zinterolum inn-latin, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-ylamino ethyl phenyl methanesulfonamide, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-yl amino ethyl phenyl methanesulfonamide, zinterolum, 2'-hydroxy-5'-l-hydroxy-2-2-methyl-1-phenyl-2-propylamino ethyl methanesulfonanilide, methanesulfonamide, n-5-2-1,1-dimethyl-2-phenylethyl amino-1-hydroxyethyl-2-hydroxyphenyl, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-yl amino ethyl phenyl methanesulfonamide, d0a0kp
CC(C)(CC1=CC=CC=C1)NCC(C2=CC(=C(C=C2)O)NS(=O)(=O)C)O
378.487
414.95
50mg
C19H26N2O4S
XJBCFFLVLOPYBV-UHFFFAOYSA-N
N-[2-hydroxy-5-[1-hydroxy-2-[(2-methyl-1-phenylpropan-2-yl)amino]ethyl]phenyl]methanesulfonamide
37990
>98%
Videos