missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF394 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25785-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.Spezifikation
ZNF394 | |
Polyclonal | |
Unconjugated | |
ZNF394 | |
zinc finger protein 394, ZKSCAN14, ZNF394 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67235, 84124 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q53GI3, Q9Z1D9 | |
Znf394 | |
ZNF394 Fusion Protein Ag22655 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |