missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
ZNF423 Polyclonal antibody specifically detects ZNF423 in Human, Mouse, Rat samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | ZNF423 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500-1:2000 |
| Zusammensetzung | PBS (pH 7.3), 50% glycerol |
| Gen-Alias | FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila) |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 220-450 of human ZNF423 (NP_055884.2). KPFKCTVCKRGFSSTSSLQSHMQAHKKNKEHLAKSEKEAKKDDFMCDYCEDTFSQTEELEKHVLTRHPQLSEKADLQCIHCPEVFVDENTLLAHIHQAHANQKHKCPMCPEQFSSVEGVYCHLDSHRQPDSSNHSVSPDPVLGSVASMSSATPDSSASVERGSTPDSTLKPLRGQKKMRDDGQGWTKVVYSCPYCSKRDFNSLAVLEIHLKTIHADKPQQSHTCQICLDSM |
| Reinigungsverfahren | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?