missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF689 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25573-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
May be involved in transcriptional regulation.Spezifikation
ZNF689 | |
Polyclonal | |
Unconjugated | |
ZNF689 | |
TIPUH1, zinc finger protein 689, ZNF689 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115509, 71131 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BKK5, Q96CS4 | |
Znf689 | |
ZNF689 Fusion Protein Ag22252 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |