Learn More
Invitrogen™ ZOOM™ IEF Fractionator Combo Kit
Artikelnummer. 10534643
Change view
Click to view available options
Packungsgröße:
5 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. unitSize
10534643 5 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 10534643 Lieferant Invitrogen™ Lieferanten-Nr. ZF10001
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen
Beinhaltet
ZOOM IEF Fractionator; One pack ZOOM disks of each pH; ZOOM reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers)
Fast, reliable method to reduce sample complexity
The ZOOM IEF Fractionator enriches low abundance proteins and increases the dynamic range of detection. Solution phase isoelectric focusing with the ZOOM IEF Fractionator provides reproducible separations in three hours. Fractionated samples are ready for further analysis by two-dimensional gel electrophoresis (2DE), one-dimensional gel electrophoresis (1DE), or two-dimensional liquid chromatography/mass spectrometry (2D LC/MS).
ZOOM IEF Fractionator
  • Built for safety, reliability, and durability
  • Includes easy-to-assemble sample chambers, in-built buffer chambers, buffer trough, removable electrode assembly, and safety lid
ZOOM Disks
  • Precast polyacrylamide gels that eliminate the need for manual preparation and minimize any chance of cross contamination
  • These immobilized buffered disks are prelabeled, disposable, and designed for single use, ensuring consistent and reproducible fractionation
  • They resolve samples into as many as seven fractions (using eight disks of specific pH) to six fractions (using six disks of specific pH) from pH 3-12; simply place each disk between the sample chambers in the ZOOM IEF Fractionator to allow separation in a specific pH range
ZOOM Reagents
  • Proteomic grade reagents, including ZOOM 2D Protein Solubilizers urea, thiourea, CHAPS, and ampholytes, that ensure optimal conditions for sample preparation
Sample Chamber and Fractionator Unit Set-up Procedure
  • The sample chamber and components fit together easily and offer a leak-proof seal
  • Simply place disks between the sample chambers in the ZOOM IEF Fractionator to allow separation in a specific pH range; for example, to fractionate proteins between pH 4.6 and 5.4, flank a sample chamber with a ZOOM Disk of pH 4.6 and one of pH 5.4
Downstream Processing by 2D Gel Electrophoresis
  • Following fractionation, the separated samples can be subjected to further analysis using narrow or broad range ZOOM Strips on the ZOOM IPGRunner™ System
  • Follow with second dimension analysis using neutral pH NuPAGE™ ZOOM Gels
Applications:
Isoelectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Spezifikation
Enthält ZOOM IEF Fractionator; One pack ZOOM disks of each pH; ZOOM reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers)