CAS RN 1000339-34-3
CAS RN 1000339-34-3
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
2-(1-(4-Bromophenyl)-1H-1,2,3-triazol-4-yl)propan-2-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.