CAS RN 1004315-81-4
CAS RN 1004315-81-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
3-(Dimethylamino)-1-(3-benzyloxyphenyl)-2S-methyl-1-propanone Dibenzoyl Tartrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(Dimethylamino)-1-(3-benzyloxyphenyl)-2S-methyl-1-propanone Dibenzoyl Tartrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-(Dimethylamino)-1-(3-benzyloxyphenyl)-2S-methyl-1-propanone Dibenzoyl Tartrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.