CAS RN 100499-89-6
CAS RN 100499-89-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Benzyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate (Benzyl Ester of Aceclofenac), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.