CAS RN 1070-62-8
CAS RN 1070-62-8
IUPAC Name:
5-ethoxy-5-oxopentanoic acid
Synonyme:
5-ethoxy-5-oxopentanoic acid
Molekulargewicht (g/mol):
160.17
Summenformel:
C7H12O4
InChi Key:
MYMNBFURSYZQBR-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCC(O)=O
1
–
1
von
1
Ergebnisse
1
Ethylwasserstoff-Glutarat, 97 %, Thermo Scientific Chemicals
CAS: 1070-62-8 Summenformel: C7H12O4 Molekulargewicht (g/mol): 160.169 MDL-Nummer: MFCD00014379 InChI-Schlüssel: MYMNBFURSYZQBR-UHFFFAOYSA-N Synonym: ethyl hydrogen glutarate,glutaric acid monoethyl ester,5-ethoxy-5-oxo-pentanoic acid,ethyl glutarate,mono,acmc-20akx9,ksc493i1d,pentanedioic acid 1-ethyl ester,pentanedioic acid monoethyl ester,5-ethoxy-5-oxidanylidene-pentanoic acid,glutaric acid monoethyl ester; monoethyl glutarate PubChem CID: 66114 IUPAC-Name: 5-Ethoxy-5-Oxopentanosäure SMILES: CCOC(=O)CCCC(=O)O