CAS RN 118021-35-5
CAS RN 118021-35-5
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MedChemExpress L-4-Oxalysine hydrochloride
MedChemExpress L-4-Oxalysine hydrochloride is a natural product isolated from the culture media of Streptomyces roseovirdofuscus in China which has shown antitumor activities.