CAS RN 1197397-26-4
IUPAC Name:
ethyl 3-(5-bromo-2-methoxypyridin-3-yl)prop-2-enoate
Synonyme:
ethyl 3-(5-bromo-2-methoxypyridin-3-yl)prop-2-enoate
Molekulargewicht (g/mol):
286.13
Summenformel:
C11H12BrNO3
InChi Key:
YEQTUBXMMANUMB-UHFFFAOYSA-N
SMILES:
CCOC(=O)C=CC1=CC(Br)=CN=C1OC
1
–
1
von
1
Ergebnisse
1
Ethyl3-(5-brom-2-methoxy-3-pyridyl)acrylat, Thermo Scientific™
CAS: 1197397-26-4 Summenformel: C11H12BrNO3 Molekulargewicht (g/mol): 286.125 MDL-Nummer: MFCD07781177 InChI-Schlüssel: YEQTUBXMMANUMB-SNAWJCMRSA-N Synonym: 3-5-bromo-2-methoxy-pyridin-3-yl-acrylic acid ethyl ester,ethyl 3-5-bromo-2-methoxypyridin-3-yl acrylate,ethyl 3-5-bromo-2-methoxy-3-pyridyl acrylate,e-ethyl 3-5-bromo-2-methoxypyridin-3-yl acrylate,3-5-bromo-2-methoxy-3-pyridyl acrylic acid ethyl ester,ethyl 2e-3-5-bromo-2-methoxy 3-pyridyl prop-2-enoate,ethyl 2e-3-5-bromo-2-methoxypyridin-3-yl prop-2-enoate PubChem CID: 42553148 IUPAC-Name: Ethyl (E)-3-(5-Brom-2-Methoxypyridin-3-yl)prop-2-enoat SMILES: CCOC(=O)C=CC1=CC(=CN=C1OC)Br