CAS RN 1309871-50-8
CAS RN 1309871-50-8
IUPAC Name:
tetrakis(triethylazanium) {[(3,4-dihydroxy-5-{2-oxo-4-[(3-phenylpropoxy)amino]-1,2-dihydropyrimidin-1-yl}oxolan-2-yl)methyl phosphonato]oxy}(phosphonatooxy)phosphinate
Synonyme:
tetrakis(TEAH) [(3,4-dihydroxy-5-{2-oxo-4-[(3-phenylpropoxy)amino]pyrimidin-1-yl}oxolan-2-yl)methyl phosphonato]oxy(phosphonatooxy)phosphinate
Molekulargewicht (g/mol):
1022.11
Summenformel:
C42H86N7O15P3
InChi Key:
ZCDZFMKRHPANNI-UHFFFAOYNA-N
SMILES:
CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC.CC[NH+](CC)CC.OC1C(O)C(OC1COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)N1C=CC(NOCCCC2=CC=CC=C2)=NC1=O