CAS RN 1314-62-1
Vanadium(V)-oxid, 99+ %, Thermo Scientific Chemicals
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 SMILES: O=[V](=O)O[V](=O)=O
Vanadium(V)-oxid, 99.6 % min., Thermo Scientific Chemicals
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 IUPAC-Name: Dioxovanadiooxy(dioxo)vanadium SMILES: O=[V](=O)O[V](=O)=O
Vanadium(V)-oxid, +98 %, Thermo Scientific Chemicals
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 SMILES: O=[V](=O)O[V](=O)=O
Vanadium(V)-oxid, +99.6 %, Thermo Scientific Chemicals
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 IUPAC-Name: Dioxovanadiooxy(dioxo)vanadium SMILES: O=[V](=O)O[V](=O)=O
Vanadium(V)-oxid, 99.2 %, Thermo Scientific Chemicals
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 SMILES: O=[V](=O)O[V](=O)=O
Vanadium(V)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 IUPAC-Name: Dioxovanadiooxy(dioxo)vanadium SMILES: O=[V](=O)O[V](=O)=O
Vanadium, Plasma-Standardlösung, Thermo Scientific Chemicals, Specpure™
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N PubChem CID: 14814 ChEBI: CHEBI:30045 IUPAC-Name: Dioxovanadiooxy(dioxo)vanadium SMILES: O=[V](=O)O[V](=O)=O
Vanadium(V)-oxid, 99.99 % (Metallbasis), Puratronic™, Thermo Scientific Chemicals
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 SMILES: O=[V](=O)O[V](=O)=O
Vanadiumpentoxid, Pure, Fisher Chemical
CAS: 1314-62-1 Summenformel: O5V2 Molekulargewicht (g/mol): 181.88 MDL-Nummer: MFCD00011457 InChI-Schlüssel: GNTDGMZSJNCJKK-UHFFFAOYSA-N Synonym: vanadium v oxide,vanadium pentoxide,divanadium pentaoxide,divanadium pentoxide,vanadic anhydride,vanadin v oxide,vanadium oxide v2o5,vanadium pentaoxide,vanadic acid anhydride PubChem CID: 14814 ChEBI: CHEBI:30045 IUPAC-Name: Dioxovanadiooxy(dioxo)vanadium SMILES: O=[V](=O)O[V](=O)=O