CAS RN 1345-25-1
IUPAC Name:
oxoiron
Synonyme:
iron oxide ferrous oxide iron(II) oxide eisen(II)-oxid
Molekulargewicht (g/mol):
71.84
Summenformel:
FeO
InChi Key:
UQSXHKLRYXJYBZ-UHFFFAOYSA-N
SMILES:
O=[Fe]
1
–
1
von
1
Ergebnisse
1
Eisen(II)-oxid, 99.5 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1345-25-1 Summenformel: FeO Molekulargewicht (g/mol): 71.844 MDL-Nummer: MFCD00016095 InChI-Schlüssel: UQSXHKLRYXJYBZ-UHFFFAOYSA-N Synonym: iron ii oxide,ferrous oxide,iron monooxide,iron monoxide,iron oxide,wustite,natural wuestite,eisenoxydul,hsdb 464,eisen ii-oxid PubChem CID: 14945 ChEBI: CHEBI:50820 IUPAC-Name: Oxoeisen SMILES: O=[Fe]