CAS RN 14620-72-5
IUPAC Name:
1,6-dimethyl hexanediimidate
Synonyme:
1,6-dimethyl hexanediimidate
Molekulargewicht (g/mol):
172.23
Summenformel:
C8H16N2O2
InChi Key:
ZLFRJHOBQVVTOJ-UHFFFAOYSA-N
SMILES:
COC(=N)CCCCC(=N)OC
1
–
1
von
1
Ergebnisse
1
Dimethyladipimidat Dihydrochlorid, 97 %, Thermo Scientific Chemicals
CAS: 14620-72-5 Summenformel: C8H18Cl2N2O2 Molekulargewicht (g/mol): 245.144 MDL-Nummer: MFCD00012573 InChI-Schlüssel: IEUUDEWWMRQUDS-UHFFFAOYSA-N Synonym: dimethyl adipimidate dihydrochloride,unii-who0f998gm,who0f998gm,adipimidic acid dimethyl ester dihydrochloride,dimethyl adipimidate hcl,dmai,acmc-209tig,bicl300,dimethyladipimidate dihydrochloride PubChem CID: 12876176 IUPAC-Name: Dimethyl-Heptanediimidat; Dihydrochlorid SMILES: COC(=N)CCCCC(=N)OC.Cl.Cl