CAS RN 14788-12-6
CAS RN 14788-12-6
IUPAC Name:
3-(dimethylamino)propanoic acid hydrochloride
Synonyme:
3-(dimethylamino)propanoic acid hydrochloride
Molekulargewicht (g/mol):
153.61
Summenformel:
C5H12ClNO2
InChi Key:
JTNKXYWGZCNBCH-UHFFFAOYSA-N
SMILES:
Cl.CN(C)CCC(O)=O
1
–
1
von
1
Ergebnisse
1
3-(Dimethylamino)propionic acid hydrochloride, TRC
CAS: 14788-12-6 Summenformel: C5H11NO2 . HCl Molekulargewicht (g/mol): 153.61 Synonym: 3-(Dimethylamino)propionic Acid Hydrochloride,3-(N,N-Dimethylamino)propionic Acid Hydrochloride,3-Dimethylaminopropanoic Acid HydrochlorideN,N-Dimethyl-beta-alanine Hydrochloride,beta-(Dimethylamino)propionic Acid Hydrochloride IUPAC-Name: 3-(dimethylamino)propanoic acid;hydrochloride SMILES: Cl.CN(C)CCC(=O)O