CAS RN 15752-05-3
CAS RN 15752-05-3
IUPAC Name:
iridium(3+) triammonium hexachloride
Synonyme:
iridium(3+) triammonium hexachloride
Molekulargewicht (g/mol):
459.03
Summenformel:
Cl6H12IrN3
InChi Key:
AIDFGYMTQWWVES-UHFFFAOYSA-K
SMILES:
[NH4+].[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3]
1
–
1
von
1
Ergebnisse
1
Ammoniumhexachloroiridat(III)-hydrat, Premion™, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 15752-05-3 Summenformel: Cl6H12IrN3 Molekulargewicht (g/mol): 459.03 MDL-Nummer: MFCD00003393 InChI-Schlüssel: AIDFGYMTQWWVES-UHFFFAOYSA-K IUPAC-Name: iridium(3+) triammonium hexachloride SMILES: [NH4+].[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3]