CAS RN 161848-60-8
CAS RN 161848-60-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MedChemExpress 5-BrUTP sodium salt
MedChemExpress 5-BrUTP sodium salt can be used to label RNA to measure the transcription.