CAS RN 166328-09-2
CAS RN 166328-09-2
IUPAC Name:
potassium [3,5-bis(trifluoromethyl)phenyl]trifluoroboranuide
Synonyme:
potassium [3,5-bis(trifluoromethyl)phenyl]trifluoroboranuide
Molekulargewicht (g/mol):
320.01
Summenformel:
C8H3BF9K
InChi Key:
WCHMTVMZBOZGCS-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
1
–
1
von
1
Ergebnisse
1
Kalium3,5-bis(trifluormethyl)phenytrifluorborat, 96 %, Thermo Scientific™
CAS: 166328-09-2 Summenformel: C8H3BF9K Molekulargewicht (g/mol): 320.006 MDL-Nummer: MFCD03701625 InChI-Schlüssel: WCHMTVMZBOZGCS-UHFFFAOYSA-N Synonym: potassium 3,5-bis trifluoromethyl phenyltrifluoroborate,potassium 3,5-bis trifluoromethyl phenyl trifluoroborate,potassium 3,5-bis trifluoromethyl phenyl trifluoroboranuide,pubchem11421,c8h3bcf9k,amtb724,potassium 3,5-bis trifluoromethyl phenytrifluoroborate,potassium 3,5-bistrifluoromethylphenyltrifluoroborate,potassium 3,5-bis trifluoromethyl phenyl-trifluoroboranuide PubChem CID: 23668091 IUPAC-Name: Kalium;[3,5-bis(trifluormethyl)phenyl]-trifluorboranuid SMILES: [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+]