CAS RN 1883548-86-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R&D Systems™ PF 06260933 dihydrochloride
A Tocris product, part of R&D Systems. MAP4K4 (HGK) inhibitor; also inhibits MINK and TNIK