CAS RN 1907-33-1
Lithium-tert.-butoxid ,2.2 M-Lösung in THF, AcroSeal™, Thermo Scientific Chemicals
CAS: 1907-33-1 Summenformel: C4H9LiO Molekulargewicht (g/mol): 80.06 MDL-Nummer: MFCD00050479 InChI-Schlüssel: LZWQNOHZMQIFBX-UHFFFAOYSA-N Synonym: lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate PubChem CID: 23664764 IUPAC-Name: Lithium; 2-Methylpropan-2-olat SMILES: [Li+].CC(C)(C)[O-]
Lithium-tert-Butoxid, 99 %, Thermo Scientific Chemicals
CAS: 1907-33-1 Summenformel: C4H9LiO Molekulargewicht (g/mol): 80.055 MDL-Nummer: MFCD00050479 InChI-Schlüssel: LZWQNOHZMQIFBX-UHFFFAOYSA-N Synonym: lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate PubChem CID: 23664764 IUPAC-Name: Lithium; 2-Methylpropan-2-olat SMILES: [Li+].CC(C)(C)[O-]
Lithium-tert-butoxid, 99%
CAS: 1907-33-1 Summenformel: C4H9LiO Molekulargewicht (g/mol): 80.06 InChI-Schlüssel: LZWQNOHZMQIFBX-UHFFFAOYSA-N Synonym: lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate PubChem CID: 23664764 IUPAC-Name: Lithium;2-Methylpropan-2-Olat SMILES: [Li+].CC(C)(C)[O-]
Lithium-tert.-Butoxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 1907-33-1 Summenformel: C4H9LiO Molekulargewicht (g/mol): 80.055 MDL-Nummer: MFCD00050479 InChI-Schlüssel: LZWQNOHZMQIFBX-UHFFFAOYSA-N Synonym: lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate PubChem CID: 23664764 IUPAC-Name: Lithium; 2-Methylpropan-2-olat SMILES: [Li+].CC(C)(C)[O-]
Lithium-tert-butoxid, Thermo Scientific Chemicals
CAS: 1907-33-1 Summenformel: C4H9LiO Molekulargewicht (g/mol): 80.05 InChI-Schlüssel: LZWQNOHZMQIFBX-UHFFFAOYSA-N Synonym: lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate PubChem CID: 23664764 IUPAC-Name: Lithium;2-Methylpropan-2-Olat SMILES: [Li+].CC(C)(C)[O-]