CAS RN 1949-55-9
CAS RN 1949-55-9
IUPAC Name:
methyl 3,5-diaminobenzoate
Synonyme:
methyl 3,5-diaminobenzoate
Molekulargewicht (g/mol):
166.18
Summenformel:
C8H10N2O2
InChi Key:
IYWLDOMAEQHKJZ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(N)=CC(N)=C1
1
–
1
von
1
Ergebnisse
1
Methyl3,5-Diaminobenzoat, 99 %, Thermo Scientific Chemicals
CAS: 1949-55-9 Summenformel: C8H10N2O2 Molekulargewicht (g/mol): 166.18 MDL-Nummer: MFCD00041433 InChI-Schlüssel: IYWLDOMAEQHKJZ-UHFFFAOYSA-N Synonym: methyl 3,5-diaminobenzenecarboxylate,3,5-diaminobenzoic acid methyl ester,methyldiaminobenzenecarboxylate,3,5-diamino-benzoic acid methyl ester,benzoic acid,3,5-diamino-, methyl ester PubChem CID: 7010431 IUPAC-Name: Methyl-3,5-diaminobenzoat SMILES: COC(=O)C1=CC(=CC(=C1)N)N