CAS RN 2070009-67-3
CAS RN 2070009-67-3
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Paricalcitol-d6, MedChemExpress
MedChemExpress Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is a drug used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.