CAS RN 2098275-86-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R&D Systems™ tri-GalNAc-C5-amine
A Tocris product, part of R&D Systems. Asialoglycoprotein receptor ligand with alkylC5 linker and amine group for onward chemistry