CAS RN 215543-92-3
CAS RN 215543-92-3
IUPAC Name:
3-{4-methyl-2-[(2-oxo-2,3-dihydro-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl}propanoic acid
Synonyme:
3-{4-methyl-2-[(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl}propanoic acid
Molekulargewicht (g/mol):
296.33
Summenformel:
C17H16N2O3
InChi Key:
JNDVEAXZWJIOKB-UHFFFAOYSA-N
SMILES:
CC1=CNC(C=C2C(=O)NC3=CC=CC=C23)=C1CCC(O)=O
1
–
5
von
5
Ergebnisse
2
SU 5402, TRC
CAS: 215543-92-3 Summenformel: C17 H16 N2 O3 Molekulargewicht (g/mol): 296.32 IUPAC-Name: 3-[4-methyl-2-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid SMILES: Cc1c[nH]c(\C=C\2/C(=O)Nc3ccccc23)c1CCC(=O)O
3
Tocris Biosciences™ SU 5402
Potent FGFR and VEGFR inhibitor
4
5
SU 5402, MedChemExpress
MedChemExpress SU 5402 is a potent multi-targeted receptor tyrosine kinase inhibitor with IC50 of 20 nM, 30 nM, and 510 nM for VEGFR2, FGFR1, and PDGFRβ, respectively.