CAS RN 2204290-85-5
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R&D Systems™ FzM1.8
A Tocris product, part of R&D Systems. Frizzled 4 allosteric agonist; exhibits biased siginaling; preserves stemness