CAS RN 23279-22-3
CAS RN 23279-22-3
IUPAC Name:
dihydrogen 2-amino-3-(3-amino-4-hydroxyphenyl)propanoic acid hydrate dichloride
Synonyme:
dihydrogen 2-amino-3-(3-amino-4-hydroxyphenyl)propanoic acid hydrate dichloride
Molekulargewicht (g/mol):
287.14
Summenformel:
C9H16Cl2N2O4
InChi Key:
ITQBVYBSXFBTJX-UHFFFAOYNA-N
SMILES:
[H+].[H+].O.[Cl-].[Cl-].NC(CC1=CC=C(O)C(N)=C1)C(O)=O
1
–
1
von
1
Ergebnisse
1
Tocris Bioscience™ 3-Aminotyrosine
Unnatural amino acid for inducing red shift in fluorescent proteins and fluorescent protein-based biosensors