CAS RN 2492-75-3
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
2-Oxohexanoic Acid, TRC
CAS: 2492-75-3 Summenformel: C6H10O3 Molekulargewicht (g/mol): 130.14 Synonym: 2-Ketocaproic Acid,2-Ketohexanoic Acid,2-Oxocaproic Acid,alpha-Ketocaproic Acid,alpha-Ketohexanoic Acid,alpha-Oxo-n-caproic Acid,alpha-Oxocaproic Acid IUPAC-Name: 2-oxohexanoic acid SMILES: CCCCC(=O)C(=O)O