CAS RN 2514-30-9
CAS RN 2514-30-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
PBIT, MedChemExpress
MedChemExpress PBIT is a specific inhibitor of the Jumonji AT-rich Interactive Domain 1 (JARID1) enzymes. PBIT inhibits JARID1B (KDM5B or PLU1) histone demethylase with an IC50 of about 3 μM . PBIT also inhibits JARID1A and JARID1C with IC50s of 6 μM and 4.9 μM, respectively.