CAS RN 2527-96-0
IUPAC Name:
methyl 5-methylfuran-2-carboxylate
Synonyme:
methyl 5-methylfuran-2-carboxylate
Molekulargewicht (g/mol):
140.14
Summenformel:
C7H8O3
InChi Key:
XBYZJUMTKHUJIY-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C)O1