CAS RN 254101-10-5
IUPAC Name:
5-(benzyloxy)-3-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyme:
5-(benzyloxy)-3-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molekulargewicht (g/mol):
337.37
Summenformel:
C17H23NO6
InChi Key:
FAFJSSKTLCNWRJ-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CC(O)=O)CC(=O)OCC1=CC=CC=C1
1
–
1
von
1
Ergebnisse
1
N-Boc-L-Beta-Glutaminsäure5-Benzylester, 95%, Thermo Scientific Chemicals
CAS: 254101-10-5 Summenformel: C17H23NO6 Molekulargewicht (g/mol): 337.372 MDL-Nummer: MFCD01862861 InChI-Schlüssel: FAFJSSKTLCNWRJ-CYBMUJFWSA-N Synonym: boc-beta-hoasp obzl-oh,boc-l-beta-glutamic acid 5-benzyl ester,boc-beta-glu obzl-oh,boc-,a-hoasp obzl-oh,r-5-benzyloxy-3-tert-butoxycarbonyl amino-5-oxopentanoic acid,3r-5-benzyloxy-3-tert-butoxycarbonyl amino-5-oxopentanoic acid,fmoc-asparaginol,boc-beta-homoaspartic acid obzl,ambotzbaa6150,boc-hoasp obzl-oh PubChem CID: 2761513 IUPAC-Name: (3R)-3-[(2-Methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentansäure SMILES: CC(C)(C)OC(=O)NC(CC(=O)O)CC(=O)OCC1=CC=CC=C1