CAS RN 2749555-66-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R&D Systems™ DX3-213B
A Tocris product, part of R&D Systems. Potent NDUFS7 antagonist